Learn More
Kingfisher Biotech Inc Swine IL-25 Recombinant Protein
Supplier: Kingfisher Biotech Inc RP1678S005
The Swine IL-25 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine IL-25 applications are for cell culture, ELISA standard, and Western Blot Control. Swine IL-25 yeast-derived recombinant protein can be purchased in multiple sizes. Swine IL-25 Specifications: (Molecular Weight: 17.1 kDa) (Amino Acid Sequence: WSQKGCPHWP NCCPSKGQNP IEEWLKQNAV LMPPLEMASP TPHPESCKAS EDGPLNSRSI APWRYELDRD LNRLPQDLYH ARCLCPHCVS LRTGSHMDPL GNSELLYHNQ TVFYRRPCPG QQGAHDSYCL EPRLYQVSLA CVCVRPRVMA (150)) (Gene ID: 100154041). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.