Learn More
Kingfisher Biotech Inc Swine OPN Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1546S100
Encompass_Preferred
The Swine Osteopontin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine Osteopontin applications are for cell culture, ELISA standard, and Western Blot Control. Swine Osteopontin yeast-derived recombinant protein can be purchased in multiple sizes. Swine Osteopontin Specifications: (Molecular Weight: 31.9 kDa) (Amino Acid Sequence: LPVKQTNSGS SEEKLLSNKY TDAVATLLKP DPSQKQTFLA PQNTISSEET DDFKQETLPSKSNESPEQTD DVDDDDDEDH VDSRDTDSEE ADHADDADRS DESHHSDESD ELVTDFPTDTPATDVTPAVP TGDPNDGRGD SVVYGLRSKS KKFRRSEAQQ LDATEEDLTS HVESEETDGTPKAILVAQRL HVASDLDSQE KDSQETSQPD DRSVETRSQE QSKEYTIKTY DGSNEHSNVIESQENPKVSQ EFHSHEDKLV PDSKSEEDKH LKLRVSHELE SASSEIN (287)) (Gene ID: n/a). For research use only. Made in the USA
Catalog No. 50-253-3164
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.