Learn More
Kingfisher Biotech Inc Swine Prolactin Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1781S100
Encompass
The Swine Prolactin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine Prolactin applications are for cell culture, ELISA standard, and Western Blot Control. Swine Prolactin yeast-derived recombinant protein can be purchased in multiple sizes. Swine Prolactin Specifications: (Molecular Weight: 23.0 kDa) (Amino Acid Sequence: LPICPSGAVN CQVSLRDLFD RAVILSHYIH NLSSEMFNEF DKRYAQGRGF ITKAINSCHT SSLSTPEDKE QAQQIHHEVL LNLILRVLRS WNDPLYHLVT EVRGMQEAPD AILSRAIEIE EQNKRLLEGM EKIVGQVHPG IKENEVYSVW SGLPSLQMAD EDTRLFAFYN LLHCLRRDSH KIDNYLKLLK CRIIYDSNC (199)) (Gene ID: 396965). For research use only. Made in the USA
Catalog No. 50-253-2777
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.