Learn More
Kingfisher Biotech Inc Swine TNF alpha Biotinylated Recombinant Protein
Supplier: Kingfisher Biotech Inc RPB1838S010
The Swine TNF alpha Biotinylated yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine TNF alpha Biotinylated applications are for cell culture. Swine TNF alpha Biotinylated yeast-derived recombinant protein can be purchased in multiple sizes. Swine TNF alpha Biotinylated Specifications: (Molecular Weight: 16.9 kDa) (Amino Acid Sequence: SSSQTSDKPVAHVVANVKAEGQLQWQSGYANALLANGVKLKDNQLVVPTDGLYLIYSQVLFRGQGCPSTNVFLTHTISRIAVSYQTKVNLLSAIKSPCQRETPEGAEAKPWYEPIYLGGVFQLEKDDRLSAEINLPDYLDFAESGQVYFGIIAL) (Gene ID: 397086). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.