Learn More
Kingfisher Biotech Inc Swine TWEAK (TNFSF12) Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP0489S100
Encompass_Preferred
The Swine TWEAK yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine TWEAK applications are for cell culture, ELISA standard, and Western Blot Control. The Swine TWEAK yeast-derived recombinant protein can be purchased in multiple sizes. Swine TWEAK Specifications: (Molecular Weight: 17.3 kDa) (Amino Acid Sequence: SAPKGRKPRA RRVIAAHYEV HPRPGQDGAQ AGVDGTVSGW EEAKINSSNP LRYDRQSGQF LVTRAGLYYL YCQVHFDEGK AVYLKLDLLV DDTLALRCLE EFSATAASSL GPQLRLCQVS GLLPLRPGSS LRIRTLPWAH LKAAPFLTYF GLFQVH (156)) (Gene ID: 100127163). For research use only. Made in the USA
Catalog No. 50-241-4093
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.