Learn More
Simport™ Scientific Swingsette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks
Catalog No. p-7116153 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-556 White
22-038-557 Pink
22-038-558 Green
22-038-559 Yellow
22-038-560 Blue
22-038-561 Peach
22-038-562 Tan
22-038-563 Gray
22-038-564 Lilac
22-038-565 Orange
22-038-566 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 22-038-556 Supplier Simport™ Scientific Supplier No. M5172T
Only null left
Add to Cart
Add to Cart
Secure specimens during processing with Swingsette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks.
  • In Quickload™ stacks
  • 45° Anterior writing area
  • Easy to assemble hinged lids
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Certifications
CE-IVD
Specifications
Quantity 2000/Pk.
Color White
Material Acetal