Learn More
Research Products International Corp Switch-Grid Tube Racks, Rack/Two Blue Grids
SDP
Supplier:  Research Products International Corp 248451
Encompass_Preferred
Switch-Grid Racks allow various tube sorting capabilities with one single base rack. Simply switch removable grids to accommodate tubes from 10mm up to 30mm. Each base holds two grids that can be mixed or matched as needed. Each rack base is supplied with two grids of the same size. Racks and grids are manufactured from a durable and chemical resistant plastic that can also be autoclaved. Grids are color coded for convenience. Rubber feet keep racks from sliding on bench top; provide secure stacking while in storage.
WARNING  This product can expose you to chemicals including Formaldehyde, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-8461
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.