Learn More
RS AMERICAS INC SWITCH ROCKER 16A 250V GRN LIT
Supplier:  RS AMERICAS INC 70226890
Encompass_Preferred
Arcolectric (Bulgin Ltd.) C1353AABG2602AW Switch, Rocker, 16A, 250V, Geen Lit 125v, Black Body, High Inrush, With Legend
Catalog No. NC2855240