Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sphergo Swivel™ Cleanroom Aluminum Base
SDP

Catalog No. 18366487 Shop All New Pig Corporation Products
Change view
Click to view available options
Disposable:
Reusable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable
18-366-487 Reusable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-487 Supplier Sphergo Supplier No. CLN375

Ergonomic swivel base for Cleanroom Aluminum Extension Pole.

  • For use with Sphergo Swivel Cleaning Tools
  • 15.75 x 4 x 1.5 in.
  • Rounded edges help prevent damage to sensitive areas such as wood trim or wooden furniture legs
  • Replaceable hook-and-loop strips
  • Light aluminum frame
  • Ideal for use in cleanrooms

Specifications

Product Type Swivel Squeegee
Color-coded No
Scent Unscented
Sterility Non-sterile
Dimensions (L x W x H) 15.75 x 4 x 1.5 in.
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.