Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sphergo Swivel™ Cleanroom Aluminum Extension Pole
SDP

Catalog No. 18366486 Shop All New Pig Corporation Products
Change view
Click to view available options
Disposable:
Reusable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable
18-366-486 Reusable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-486 Supplier Sphergo Supplier No. CLN374
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ergonomically curved extension pole handle that swivels for cleanroom mop.

  • For use with Sphergo Swivel Cleaning Tools
  • 46 x 3 x 3 in.
  • Curved pole and swivel ball help position the mopping tool and body in a less stressful posture to reduce repetitive strain injuries
  • Ideal for use in cleanrooms
  • Design provides easier access into corners and along baseboards

Specifications

Product Type Mopping System
Color-coded No
Handle Material Aluminum
Handle Type Extension
Scent Unscented
Sterility Non-sterile
Dimensions (L x W x H) 46 x 3 x 3 in.
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.