Learn More
Krayden Dow Sylgard 184 Silicone Elastometer Kit. Clear, (1.1LB)
Supplier: Krayden DC4019862
DC 184 Sylgard 0.5KG 1.1LB kit. Dow Corning 184 Silicone Elastomer. Transparent encapsulant with good flame resistance, flowable, room temperature and heat cure. High tensile strength. 2 part kit, clear.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.