Learn More
MyBioSource Human Symmetric Dimethyl Arginine (SDMA) ELISA Kit
SDP
Supplier:  MyBioSource MBS1677615X96STRIPWELL
Encompass
The MyBioSource Human ELISA Kit quantifies SDMA in various human biological fluids using a 96-well sandwich ELISA technique. It includes all necessary reagents and takes about 2 hours for detection within a 0.156-10 ng/mL range. This kit is for research use only.
Catalog No. NC2210594
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.