Learn More
MyBioSource Symmetric Dimethyl ELISA Kit, 96 Wells
Supplier: MyBioSource MBS723789296STRIPWELLS
The MyBioSource Symmetric Dimethyl ELISA Kit is a 96-well strip plate kit used for the quantitative measurement of symmetric dimethylarginine (SDMA) in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. The kit uses a double-antibody sandwich ELISA technique to detect SDMA levels with high sensitivity and excellent specificity. The kit includes a microtiter plate pre-coated with an antibody specific for SDMA, a biotin conjugated SDMA antibody, Avidin conjugated Horseradish Peroxidase (HRP), standards, sample diluent and wash buffer. The kit has a detection range of 78-5000 pg/mL SDMA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.