Learn More
Waters Corp Symmetry C8 Column, 100Å, 3.5 µm, 4.6 mm X 100 mm, 1/pk
Supplier:  Waters Corp WAT066204
Encompass_Preferred
Symmetry C8 columns feature monofunctionally bonded C8 ligands on a high purity base-deactived silica. Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
Catalog No. 50-785-866
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.