Learn More
Waters Corp Symmetry C4 Column, 300Å, 5 µm, 3.9 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186000286
Encompass_Preferred
Symmetry columns continues to set the standard for HPLC drug assays, giving you confidence in the long-term compliance of your HPLC methods. No need to compromise on peak shape, selectivity, column-to-column and batch-to-batch reproducibility, or other critical performance characteristics. Available in a variety of particle sizes, chemistries and dimensions.
Catalog No. 50-780-173
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.