Learn More
Waters Corp Symmetry Shield RP8 Column, 100Å, 3.5 µm, 4.6 mm X 150 mm, 1/pk
Supplier:  Waters Corp WAT094269
Encompass_Preferred
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP8 column significantly improves peak shape and resolution.
Catalog No. 50-819-122
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.