Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

syn-2-Nitrobenzaldoxime 85.0+%, TCI America™
SDP

Catalog No. N048625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N048625G 25 g
N04865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N048625G Supplier TCI America Supplier No. N048625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4836-00-4
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.14
MDL Number MFCD00007187
InChI Key IHMGDCCTWRRUDX-UHFFFAOYSA-N
PubChem CID 7332808
IUPAC Name N-[(2-nitrophenyl)methylidene]hydroxylamine
SMILES ON=CC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 95°C
Color Yellow
Quantity 25 g
Formula Weight 166.14
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material syn-2-Nitrobenzaldoxime [Deprotecting Agent]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.