Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC Quantitative Synthetic Avian Influenza Virus (H7N7) RNA

Supplier:  ATCC VR3438SD

Encompass

Quantitative Synthetic Avian Influenza Virus (H7N7) RNA is a synthetically derived RNA standard intended for development, verification, and validation of molecular assays targeting H7N7. For laboratory research use only; not for diagnostic, therapeutic, or consumption purposes.

  • Synthetic RNA standard, frozen.
  • Unit size: 100 μL.
  • Concentration: Construct A: ≥ 1 x 10^5 to 1 x 10^6 copies/μL; Construct B: ≥ 1 x 10^5 to 1 x 10^6 copies/μL.
  • Intended for assay development, verification, validation, and monitoring assay performance.
  • Storage: -70°C or colder.
  • Technical data sheet and sequence analysis available to customers upon purchase.

Catalog No. NC3760738


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.