Learn More
Invitrogen™ Synapsin 1 Polyclonal Antibody

Description
This antibody has little or no activity against Synapsin II. Storage and reconstitution: when stored at -20°C or below, the lyophilized antibody should retain full activity for over one year. Reconstitute using 50 μL PBS, pH 7.4, to yield a 0.2 mg/mL solution. If storing at 0-4°C, add sodium azide to a final concentration of 2mM. For longer storage, divide the solution into aliquots and freeze at -20°C, avoiding freeze/thaw cycles.
Specifications
Specifications
| Antigen | Synapsin 1 |
| Applications | Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 5mM Ammonium bicarbonate with no preservative |
| Gene | SYN1 |
| Gene Accession No. | O88935, P09951, P17599, P17600 |
| Gene Alias | Brain protein 4.1; SYN1; Syn-1; SYN1a; SYN1b; Syn1-S; synapsin 1; synapsin I; synapsin Ia; synapsin Ib; synapsin-1; SYNI; truncated synapsin 1-S |
| Gene Symbols | SYN1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.