Learn More
Invitrogen™ Synapsin 1 Polyclonal Antibody

Description
The antibody should be reconstituted in 50 uL phosphate buffered saline (PBS: 137 mM NaCl, 7.5 mM Na2HPO4, 2.7 mM KCl, 1.5 mM KH2PO4, pH 7.4) before use. After reconstitution the antibody should be aliquot and stored at -20C.
Specifications
Specifications
| Antigen | Synapsin 1 |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. not determined |
| Conjugate | Unconjugated |
| Formulation | 5mM Ammonium bicarbonate with no preservative |
| Gene | SYN1 |
| Gene Accession No. | O88935, P09951, P17599, P17600 |
| Gene Alias | Brain protein 4.1; SYN1; Syn-1; SYN1a; SYN1b; Syn1-S; synapsin 1; synapsin I; synapsin Ia; synapsin Ib; synapsin-1; SYNI; truncated synapsin 1-S |
| Gene Symbols | SYN1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.