Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptojanin 1 Monoclonal Antibody (AC1)
GREENER_CHOICE

Catalog No. 01670186 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-186 Supplier Invitrogen™ Supplier No. MA3936
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3-936 detects Synaptojanin 1 from rat and human samples. MA3-936 has been successfully used in Western blotting, IHC (frozen), IP and ICC/IF procedures. By Western blot MA3-936 detects a band at 170 kDa representing Synaptojanin 1. The MA3-936 immunogen is proline rich domain of a fusion protein comprising aa 1156-1286 of rat synaptojanin 1.

Synaptojanin 1 is a phosphoinositide phosphatase that regulates levels of membrane phosphatidylinositol-4,5-bisphosphate. As such, expression of this enzyme may affect synaptic transmission and membrane trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptojanin 1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone AC1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites, PBS with 0.05% sodium azide
Gene SYNJ1
Gene Accession No. O43426, Q62910
Gene Alias A930006D20Rik; AA675315; inositol 5'-phosphatase (synaptojanin 1); inositol polyphosphate-5-phosphatase G; INPP5G; KIAA0910; mKIAA0910; PARK20; phosphoinositide 5-phosphatase; synaptic inositol 1,4,5-trisphosphate 5-phosphatase 1; synaptic inositol-1; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 1; synaptojanin 1; synaptojanin-1; synaptojanin-1, polyphosphoinositide phosphatase; Synj1
Gene Symbols SYNJ1
Host Species Mouse
Immunogen Proline rich domain of a fusion protein comprising amino acids 1156-1286 of rat synaptojanin 1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85238, 8867
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.