Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptojanin 1 Recombinant Superclonal™ Antibody

Catalog No. 711821
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-182-1 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-182-1 Supplier Invitrogen™ Supplier No. 711821
Add to Cart
Add to Cart

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Monkey, Dog, Horse Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

Synaptojanin 1 is a phosphoinositide phosphatase that regulates levels of membrane phosphatidylinositol-4,5-bisphosphate. As such, expression of this enzyme may affect synaptic transmission and membrane trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptojanin 1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Recombinant Superclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene SYNJ1
Gene Accession No. O43426, Q62910, Q8CHC4
Gene Alias A930006D20Rik; AA675315; inositol 5'-phosphatase (synaptojanin 1); inositol polyphosphate-5-phosphatase G; INPP5G; KIAA0910; mKIAA0910; PARK20; phosphoinositide 5-phosphatase; synaptic inositol 1,4,5-trisphosphate 5-phosphatase 1; synaptic inositol-1; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 1; synaptojanin 1; synaptojanin-1; synaptojanin-1, polyphosphoinositide phosphatase; Synj1
Gene Symbols SYNJ1
Host Species Rabbit
Immunogen Protein corresponding to human Synaptojanin 1 [aa1156-aa1286].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104015, 85238, 8867
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.