Learn More
Cusabio Technology LLC Human synaptophysin (SYP) ELISA kit
Supplier:  Cusabio Technology LLC CSBE17406H
Encompass_Preferred
This Human SYP ELISA Kit was designed for the quantitative measurement of Human SYP protein in serum, plasma, tissue homogenates, cell lysates. It is a Sandwich ELISA kit, its detection range is 62.5 pg/mL-4000 pg/mL and the sensitivity is 15.6 pg/mL. One kit contains 96 wells.
Catalog No. NC2952974