Learn More
Biocare Medical, LLC Synaptophysin Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM371AA
Encompass_Preferred
Synaptophysin antibody [clone 27G12] is a reliable marker for neuroendocrine and neuronal neoplasms, as well as their normal counterparts. Synaptophysin has a higher degree of specificity than NSE, but is often used in conjunction with CD56, NSE and Chromogranin. This new clone is very specific and provides sharper staining when compared to other rabbit synaptophysin antibodies. In-house studies, reference laboratories, European institutions and NordiQC have validated this antibody as a first line screener for neuroendocrine tumors.
Catalog No. 50-831-43
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.