Learn More
Carbosynth LLC. Mouse monoclonal Synaptopodin antibody
SDP
Supplier:  Carbosynth LLC. 10RS125A
Encompass_Preferred
Synaptopodin is a protein that in humans is encoded by the SYNPO gene. Synaptopodin is an actin-associated protein that may play a role in actin-based cell shape and motility. The name synaptopodin derives from the protein's associations with postsynaptic densities and dendritic spines and with renal podocytes.
Catalog No. 50-125-2778
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.