Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syndecan 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 363100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-310-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-310-0 Supplier Invitrogen™ Supplier No. 363100
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Syndecan 4 Polyclonal Antibody for ELISA, IHC (P), Western Blot

Syndecans are type I integral membrane proteoglycans that are involved in cell-extracellular matrix adhesion and growth factor binding. Syndecan 1 is a matrix receptor which binds to Collagens, Fibronectin and Thrombospondin. Syndecan 1 and Syndecan 3 interact with MK (midkine), a growth/differentiation factor involved in embryogenesis. Syndecan 2 is highly expressed at areas of high morphogenetic activity, such as epithelial-mesenchymal interfaces and the prechondrogenic and preosteogenic mesenchymal condensations. Syndecan 4 functions cooperatively with integrins in the processes of cell spreading and focal adhesion assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syndecan 4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Sdc4
Gene Accession No. P31431
Gene Alias AA959608; Amphiglycan; AW108331; MGC22217; RATRYUDOCA; RYUDOCA; ryudocan; ryudocan amphiglycan; ryudocan core protein; Ryudocan/syndecan 4; S4; SDC4; Synd4; syndecan 4; syndecan 4 (amphiglycan, ryudocan); syndecan proteoglycan 4; syndecan-4
Gene Symbols Sdc4
Host Species Rabbit
Immunogen Synthetic peptide derived from the middle region of the human Syndecan-4 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6385
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.