Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Synergy H1 Hybrid Multi-Mode Reader
GSA_VAGREENER_CHOICE

Catalog No. BTH1FG Shop All Agilent Technologies Products
Change view
Click to view available options
Model:
H1F
H1FG
H1M
H1M2
H1M2F
H1M2FG
H1M2G
H1MF
H1MFG
H1MG
Incubator Temperature:
4-Zone Incubation to 45°C
4-Zone Incubation to 70°C
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Incubator Temperature
BTH1FG H1FG 4-Zone Incubation to 45°C
11120531 H1F 4-Zone Incubation to 45°C
11-120-533 H1M 4-Zone Incubation to 45°C
11-120-535 H1MF 4-Zone Incubation to 45°C
BTH1MG H1MG 4-Zone Incubation to 45°C
BTH1MFG H1MFG 4-Zone Incubation to 45°C
BTH1M2 H1M2 4-Zone Incubation to 70°C
BTH1M2F H1M2F 4-Zone Incubation to 70°C
BTH1M2FG H1M2FG 4-Zone Incubation to 70°C
BTH1M2G H1M2G 4-Zone Incubation to 70°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. BTH1FG Supplier Agilent BioTek Supplier No. SH1FGSN

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements.

This item is not returnable. View return policy

BioTek Synergy H1 is a modular multi-mode microplate reader, with monochromator-based optics and filter-based optics. For Research Use Only. Not for use in diagnostic procedures.

BioTek Synergy H1 is a modular multi-mode microplate reader, with monochromator-based optics and filter-based optics. The patented Hybrid Technology offers flexibility and sensitivity across a broad range of applications. The modular platform allows upgrading to expand functionality as your laboratory workflows change

Synergy H1 offers continuously variable bandwidth monochromators for fluorescence excitation and emission wavelength selection. Fluorescence bandwidth can be set between 9 and 50 nm, in 1 nm increments, allowing users to fully optimize reader settings to drive the best assay performance compared to fixed bandwidth systems.

Order Info

Gas module must be purchased separately (BT1210007 or BT12310008)

Certifications

CE marked

Compliance

RoHS Compliant

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Absorbance Range Absorbance: 230 to 999 nm (1 nm increments)
Accuracy Absorbance: ±2 nm; Dispenser: ±1 μL or 2%
Certifications/Compliance CE and TUV marked. RoHS compliant.
CO2 Concentration Range 0 to 20%
Depth (Metric) 46.4 cm
Compatibility BioStack and Third Party Automation Compatible
Depth (English) 18.3 in.
Incubator Temperature 4-Zone Incubation to 45°C
Measurement Speed 11 sec. (96 wells), 22 sec. (384 wells)
Shaking Linear, Orbital, Double Orbital
Dynamic Range 0 to 4 OD
Dispense Volume 5 to 1000 μL, 1 μL increments
Height (English) 13 in.
Frequency 50/60 Hz
Height (Metric) 33 cm
Includes Compatible with gas controller; Full-light luminescence; Filter/dichroic fluorescence; Fluorescence polarization; Time resolved fluorescence; Filtered luminescence
Measurement Technology Proprietary Hybrid Technology
Model H1FG
No. of Dispensers 2 Syringe Pumps
Product Type Multi-Mode Microplate Reader
Power Consumption 130 W max.
Resolution ±0.1% (CO2 and O2)
Sensitivity 2.5 to 4 pM monochromators, 0.25 pM filters fluorescence, 20 amol ATP monochromator, 10 amol ATP filter luminescence, 1.2 mP fluorescence polarization, Europium 40 fM with filters, 1200 fM with monos time resolved fluorescence
Wavelength Range Fluorescence Intensity Monochromator: 250 to 700 nm (900 nm option), Fluorescence Polarization: 280 to 700 nm (850 nm option)
Weight (English) <55 lb.
Weight (Metric) 25 kg
Width (English) 14.8 in.
Width (Metric) 37.5 cm
Plate Type 6 to 384-well Plates, Take3 Micro-volume Plates
Detector Type Photodiode, PMT
Software Type Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Light Source Xenon Flash
Reading Options Endpoint, Kinetic, Spectral Scanning, Well Area Scanning
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.