Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Synergy HTX Multi-Mode Microplate Reader
GSA_VA

Catalog No. bts1la Shop All Agilent Technologies Inc. Products
Change view
Click to view available options
Model:
S1A
S1L
S1LA
S1LF
S1LFA
S1LFTA
Includes:
Absorbance, Incubation, Shaking
Absorbance, Luminescence, Fluorescence (Top/Bottom), Alpha, Incubation, Shaking
Absorbance, Luminescence, Fluorescence (Top/Bottom), Time-resolved Fluorescence (TRF - Secondary Mode), Alpha, Incubation, Shaking
Absorbance, Luminescence, Incubation, Shaking
Luminescence, Fluorescence (Top/Bottom), Alpha, Incubation, Shaking
Luminescence, Incubation, Shaking
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes
BTS1LA S1LA Absorbance, Luminescence, Incubation, Shaking
BTS1LFA S1LFA Absorbance, Luminescence, Fluorescence (Top/Bottom), Alpha, Incubation, Shaking
BTS1A S1A Absorbance, Incubation, Shaking
BTS1L S1L Luminescence, Incubation, Shaking
BTS1LF S1LF Luminescence, Fluorescence (Top/Bottom), Alpha, Incubation, Shaking
BTS1LFTA S1LFTA Absorbance, Luminescence, Fluorescence (Top/Bottom), Time-resolved Fluorescence (TRF - Secondary Mode), Alpha, Incubation, Shaking
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. BTS1LA Supplier Agilent BioTek Supplier No. S1LASN

The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. For Research Use Only. Not for use in diagnostic procedures.

The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. The unique dual optics design provides superior performance for UV-Vis absorbance, fluorescence, luminescence, and AlphaScreen/AlphaLISA workflows

With incubation and shaking, plus a dual reagent injector module, the system will meet all your assay requirements now and in the future. Synergy HTX is controlled by the easy-to-use, yet powerful, Gen5 software for data collection, analysis, exporting, and reporting.

Order Info

Optional Accessories: Dual Reagent Dispenser Module, Gen5 Secure (for 21 CFR Part 11 Compliance), Fluorescence Test Plate, Absorbance Test Plate, Luminescence Test Plate, Patented BioCell 1 cm quartz vessel, Product Qualification Package, Take3/Take3 Trio.

Certifications

CE and TUV marked

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Absorbance Range 200 to 999 nm, 1 nm increments
Accuracy Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2%
Certifications/Compliance CE and TUV marked. RoHS compliant.
Depth (Metric) 38 cm
Compatibility BioStack and Third Party Automation Compatible
Depth (English) 15 in.
Incubator Temperature 4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C)
Measurement Speed 31 sec. (96 wells), 80 sec. (384 wells)
Shaking Linear, Orbital
Dynamic Range Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades
Height (English) 10 in.
Frequency 50/60 Hz
Height (Metric) 25.4 cm
Includes Absorbance, Luminescence, Incubation, Shaking
Model S1LA
No. of Dispensers 2 Syringe Pumps
Product Type Multi-Mode Microplate Reader
USB Connections 2
Voltage 100/240 V
Weight (English) 40 lb.
Weight (Metric) 18 kg
Width (English) 16 in.
Width (Metric) 40.6 cm
Plate Type 6- to 384-well Plates
Detector Type Photodiode, PMT
Software Type Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Light Source Xenon Flash, Tungsten Halogen (Alpha Detection/Fluorescence Intensity)
Reading Options End Point, Kinetic, Spectral Scanning, Well Area Scanning
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.