Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader
GSA_VAGREENER_CHOICE

Catalog No. btneo2sm Shop All Agilent Technologies Inc. Products
Change view
Click to view available options
Model:
NEO2
NEO2ALPHA
NEO2ALPHAB
NEO2ALPHABT
NEO2ALPHAT
NEO2B
NEO2M
NEO2MALPHA
NEO2MALPHAB
NEO2MALPHABT
NEO2MALPHAT
NEO2MB
NEO2MBT
NEO2MON
NEO2MT
NEO2S
NEO2SALPHA
NEO2SALPHAB
NEO2SB
NEO2SM
NEO2SMALPHA
NEO2SMALPHAB
NEO2SMB
NEO2T
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
BTNEO2SM NEO2SM
BTNEO2 NEO2
BTNEO2M NEO2M
BTNEO2MB NEO2MB
BTNEO2S NEO2S
BTNEO2SMB NEO2SMB
BTNEO2MBT NEO2MBT
BTNEO2MON NEO2MON
BTNEO2MT NEO2MT
BTNEO2T NEO2T
NEO2ALPHABT NEO2ALPHABT
NEO2ALPHAT NEO2ALPHAT
NEO2MALPHAT NEO2MALPHAT
NE2MALPHABT NEO2MALPHABT
NE2SMALPHAB NEO2SMALPHAB
NEO2MALPHA NEO2MALPHA
NEO2MALPHAB NEO2MALPHAB
NEO2SALPHA NEO2SALPHA
NEO2SMALPHA NEO2SMALPHA
BTNEO2ALPHA NEO2ALPHA
BTNEO2B NEO2B
BTNEO2SB NEO2SB
NEO2ALPHAB NEO2ALPHAB
NEO2SALPHAB NEO2SALPHAB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. BTNEO2SM Supplier Agilent BioTek Supplier No. N2SMSN

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements.

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For Research Use Only. Not for use in diagnostic procedures.

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. Variable bandwidth quad monochromators, sensitive high transmission filter-based optics, laser TRF, and up to 4 PMTs provide ultrafast measurements.

Advanced environmental controls, including CO2/O2 control, incubation to 70°C, and variable shaking support live cell assays. BioStack Neo delivers walk-away automation, high throughput, barcode-labeled filter cubes to streamline workflows and limit errors. Powerful Gen5 software provides complete reader control, powerful data analysis, and LIMS integration.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Absorbance Range 230 to 999 nm, 1 nm increments
Accuracy Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD
Certifications/Compliance CE and TUV marked. RoHS compliant.
CO2 Concentration Range 0 to 20%
Depth (Metric) 52.5 cm
Depth (English) 20.7 in.
Incubator Temperature ±0.2°C at 37°C
Shaking Linear, Orbital, Double Orbital
Dynamic Range 0 to 4 OD
Dispense Volume 5 to 1000 μL, 1 μL increments
For Use With (Application) HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP
Height (English) 16.1 in.
Height (Metric) 41 cm
Linearity ≤1% from 0 to 3 OD
Includes Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM
Model NEO2SM
Product Type Multi-Mode Microplate Reader
Power Consumption 250 W max.
Resolution 0.0001 OD
Weight (English) 78 lb.
Weight (Metric) 35 kg
Width (English) 15.4 in.
Width (Metric) 39 cm
Detector Type Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option)
Software Type Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Light Source Xenon Flash
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.