Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synoptics™ Syngene™ Blue Light Converter Screen
GREENER_CHOICESDP

Catalog No. 01257180 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Size:
25 x 30cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01-257-180 25 x 30cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-180 Supplier Synoptics™ Supplier No. CONVERTBLUE2530
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Convert UV to blue light for imaging of Safe Dyes - use with Uv transilluminators

  • UV-to-blue light converter screen
  • Area 25 x 30cm

convertor, uv to blue, blue screen

Specifications

For Use With (Application) Safe dyes
Product Type Convertor Screen
Quantity 1Each
Color Blue
For Use With (Equipment) Syngene Gbox
Size 25 x 30cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.