Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synoptics™ Gel Imaging DigiGenius
GREENER_CHOICESDP

Catalog No. 01257157 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Display Type:
None
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
01-257-157 None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-157 Supplier Synoptics™ Supplier No. DG1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DigiGenius is a digital camera with hood UV filter. One copy of GeneTools. Transilluminator converter not supplied. Standard Warranty 1 year. PC required (not included).

The DigiGenius offers reproducible, high resolution digital images for gel documentation. The system is easy to use and allows you to capture and view images directly on your PC or MAC. DigiGenius gel documentation system is ideal if you have a limited budget because you can purchase the basic system and then add a UV transilluminator, white light table or conversion screen, safety hood, mini darkroom and printer, as well as Syngene's industry leading GeneTools analysis software, as the need arises.
  • Electrophoresis gels - Ethidium Bromide, Coomassie Blue, Silver Stain, SYBR™ Gold, SYBR™ Green, SYBR™ Safe, GelStar™, SYPRO™ Red, SYPRO™ Ruby, SYPRO™ Orange, Fluorescein, Rhodamine Red™, Texas Red™, Pro-Q™ Diamond , Deep Purple™
  • Agar plates
  • Cells in flasks
  • Autoradiographs
  • Membranes
  • Spots and slot blots
  • Microtitre plates
  • Macro-arrays
  • Low cost gel documentation system
  • Digital camera - high quality images with precise band separation
  • Budget system with analysis software - cost effective method of upgrading as fluorescent applications change
  • Interface to PC or Mac - full control of camera set-up and image capture

Note: A transilluminator needs to be purchased separately.

Specifications

Voltage 115 V
Depth (English) 11 in.
Depth (Metric) 210 mm
Filter Type UV
Height (English) 8.2 in.
Pixel Size 18 MP
Width (English) 14.6 in.
Height (Metric) 370 mm
Width (Metric) 370 mm
Camera Type 24 Bit Color
For Use With (Application) Gel Imaging
Product Line Syngene
Product Type Gel Documentation System
Display Type None
Frequency 50 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.