Learn More
Synoptics™ HILED Lighting Option
GREENER_CHOICESDP
Catalog No. 01257277 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Wavelength:
Blue, green, red, infrared
Catalog No. Wavelength
01-257-277 Blue, green, red, infrared
1 options
Catalog No. 01-257-277 Supplier Synoptics™ Supplier No. HILEDRGBIR
Add to Cart
Add to Cart
Request a Quote
HILED option for illumination of multiplex fluorescence blots - for use in Syngene Gbox
  • HILED Lighting option for GBOX
  • Lighting units: Red, Green, Blue and Infra-Red
  • Includes gantry and electronics
HILED, Multiplex. Fluorescence, westerns
Specifications
Product Line Syngene
For Use With (Application) Fluorescence westerns
Product Type HILEDS
Quantity 1Each
For Use With (Equipment) Syngene Gbox
Wavelength Blue, green, red, infrared