Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNJ2BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551471 Supplier Invitrogen™ Supplier No. PA551471
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NGRVDYLVTE EEINLTRGPS GLGFNIVGGT DQQYVSNDSG IYVSRIKENG AAALDGRLQE GDKILSVNGQ DLKNLLHQDA VDLFRNAGYA VSLRVQHRLQ VQNGPIGHRG EG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

Regulates endocytosis of activin type 2 receptor kinases through the Ral/RALBP1-dependent pathway and may be involved in suppression of activin-induced signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNJ2BP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYNJ2BP
Gene Accession No. P57105, Q9D6K5
Gene Alias AA409442; activin receptor interacting protein 2; activin receptor interacting protein 5; activin receptor-interacting protein 2; activin receptor-interacting protein 2a; activin receptor-interacting protein 4; ActRIP4; Arip2; ARIP2a; D12Wsu118e; Mitochondrial outer membrane protein 25; NPW16; Omp25; outer membrane protein; outer membrane protein 25; synaptojanin 2 binding protein; synaptojanin-2-binding protein; SYNJ2BP
Gene Symbols SYNJ2BP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYNJ2BP. Recombinant protein control fragment (Product #RP-88909).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24071, 55333
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.