Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNPO2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596246 Supplier Invitrogen™ Supplier No. PA596246
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HVTPKTPPPM APKTPPPMTP KTPPPVAPKP PSRGLLDGLV NGAASSAGIP EPPRLQGRGG ELFAKRQSRA DRYVVEGTPG PGLGPRPRSP SPTPSLPPSW K; Positive Samples: Mouse heart; Cellular Location: Cytoplasm, cytoskeleton.

SYNPO2L was initially identified as a novel heart-enriched gene that encodes a cytoskeletal protein highly expressed in the Z-disc of heart and skeletal muscle, associates with actin and interacts with a-actinin. It is a member of the synaptopodin family, sharing greatest homology with Synaptopodin 2. Recent studies have shown that SYNPO2L, while primarily localized to the sarcomere, can also translocate to the nucleus. A knockdown of SYNPO2L in zebrafish resulted in aberrant cardiac and skeletal muscle development and function, suggesting that it is a critical component of the sarcomere and plays an important role in muscle development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNPO2L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SYNPO2L
Gene Accession No. Q8BWB1
Gene Alias 1110054M18Rik; Chap; RGD1565434; synaptopodin 2 like; synaptopodin 2-like; synaptopodin 2-like protein; SYNPO2L
Gene Symbols SYNPO2L
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 700-800 of human SYNPO2L (NP_0011076051).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68760
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.