Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 6 Mouse anti-Human, Mouse, Rat, Clone: 2B6G4, Proteintech
SDP

Catalog No. 501736213 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6213 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6213 Supplier Proteintech Group Inc Supplier No. 600591IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STX6 (Syntaxin 6) is a Protein Coding gene. Diseases associated with STX6 include Supranuclear Palsy, Progressive, 1. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Vesicle-mediated transport. Gene Ontology (GO) annotations related to this gene include syntaxin binding and SNAP receptor activity.

Specifications

Antigen Syntaxin 6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2B6G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene STX6
Gene Accession No. O43752, Q63635, Q9JKK1
Gene Alias STX6, syntaxin 6
Gene Symbols Stx6
Host Species Mouse
Immunogen Syntaxin 6 Fusion Protein Ag1284
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10228, 58244, 60562
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.