Learn More
Cayman Chemical SynthetIc a-GuaIen 5mg
SDP
Supplier:  Cayman Chemical 346865 MG
Encompass_Preferred
A glycine-site selective NMDA receptor antagonist selectively binds to the NMDA receptor glycine-binding site (IC50 0 56 UM) over the NMDA- quisqualate- and kainate-binding sites (IC50s 169 153 and 1000 UM respectively) induces antidepressant-like activity in a variety of behavioral tests in mice at 0 1 and 1 mg/kg
Catalog No. 50-291-0252
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.