Learn More
R&D Systems™ Synthetic Human Ubiquitin Vinyl Sulfone Protein, CF
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Accession Number | P0CG47.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a solution in MES and DMSO. |
| Gene ID (Entrez) | 6233 |
| Quantity | 50 μg |
| Source | Chemically Synthesized Ubiquitin protein Met1 - Gly75 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Gene Alias | RPS27A, UBA52, UBB, UBB ubiquitin B, UBC |
| Conjugate | Vinyl Sulfone |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.