Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthetic Perspiraton B Ammonium Hydroxide and Sodium Chloride in Water, Reagents
Synthetic Perspiraton B Ammonium Hydroxide + Sodium Chloride in Water, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CS500300100A
Quantity | 100 mL |
---|
Chemical Identifiers
7732-18-5, 7647-14-5, 1336-21-6 | |
18.02 | |
Artificial sweat | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
Colorless | |
100°C | |
Artificial sweat | |
O | |
18.02 | |
Poly Bottle | |
100 mL | |
Liquid |
7732-18-5, 7647-14-5, 1336-21-6 | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1.0026 | |
Non-sterile | |
Synthetic Perspiraton B Ammonium Hydroxide + Sodium Chloride in Water |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction