Learn More
Cayman Chemical SynthetIc TectrIdIn 25mg
SDP
Supplier:  Cayman Chemical 3396925 MG
Encompass_Preferred
An inhibitor of GlyT1 (IC50 3 nM) selective for GlyT1 over GlyT2 (IC50 10 UM) as well as over NMDA receptors and the inhibitory glycine receptor at 100 UM increases prefrontal cortical levels of glycine in rats at 10 mg/kg reverses MK-801-induced persistent latent inhibition in a mouse model of schizophrenia
Catalog No. 50-294-9839
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.