Learn More
Cayman Chemical galacto-DapaglIflozIn 5mg
Supplier: Cayman Chemical 159525 MG
A natural broad spectrum Gram-positive antibiotic that inhibits two key enzymes in bacterial fatty acid synthesisBketoacyl-ACP synthases II and III (FabF and FabH respectively) IC50 1.95 and 3.91 g/ml respectively does not exhibit cross-resistance to antibiotic resistant bacterial strains
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.