Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Micro Funnel
GREENER_CHOICESDP

Catalog No. 31502125 Shop All Kemtech Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-502-125 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-502-125 Supplier Synthware™ Supplier No. F740027
Add to Cart
Add to Cart

For use with microscale applications

Specifications

Color Clear
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Type Micro Funnel
Diameter (Metric)Top 27 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.