Learn More
Synthware™ Adapters with Ball Socket
GREENER_CHOICESDP
Catalog No. 31500521 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
Top: 35/20, Bottom: 14/20
Top: 35/20, Bottom: 24/40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-521 Top: 35/20, Bottom: 14/20
31-500-522 Top: 35/20, Bottom: 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-500-521 Supplier Synthware™ Supplier No. A663514
Add to Cart
Add to Cart
With top spherical socket joint and lower standard taper inner joint
Synthware combination adapting connector with a top spherical socket joint and a lower standard taper inner joint
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Adapter with ball socket
Quantity 1
Standard Taper Joint No. Top: 35/20, Bottom: 14/20