Learn More
Synthware™ Addition Funnels with Pressure Equalizing Arm and 1:5 PTFE Stopcock
Shop All Kemtech Products
Description
- Top outer and lower inner standard taper joints of same size
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Joint Number | 19/22 |
| Product Type | Pressure-equalizing Funnel |