Description
- With pressure equalizing arm and high vacuum valve
- Top outer and lower inner standard taper joints of same size
Specifications
Specifications
| Diameter (Metric)Top | Valve orifice: 0-4mm |
| Capacity (Metric) | 60 mL |
| Joint Number | 19/22 |
| Product Type | Pressure-equalizing Funnel with High Vacuum Valve |