Learn More
Synthware™ Bushing Adapters
GREENER_CHOICESDP
Catalog No. 31500502 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
Top Outer: 14/20, Bottom Inner: 19/22
Top Outer: 14/35, Bottom Inner: 19/38
Top Outer: 14/35, Bottom Inner: 24/40
Top Outer: 19/38, Bottom Inner: 24/40
Top Outer: 24/40, Bottom Inner: 34/45
Top Outer: 24/40, Bottom Inner: 45/50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-502 Top Outer: 14/35, Bottom Inner: 24/40
31-500-500 Top Outer: 14/20, Bottom Inner: 19/22
31-500-501 Top Outer: 14/35, Bottom Inner: 19/38
31-500-503 Top Outer: 19/38, Bottom Inner: 24/40
31-500-504 Top Outer: 24/40, Bottom Inner: 34/45
31-500-505 Top Outer: 24/40, Bottom Inner: 45/50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31-500-502 Supplier Synthware™ Supplier No. A793524
Add to Cart
Add to Cart
For compact assemblies of dissimilar joints
Synthware bushing adapters allow for compact assemblies of dissimilar standard taper joints.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Bushing adapter
Quantity 1
Standard Taper Joint No. Top Outer: 14/35, Bottom Inner: 24/40