Learn More
Synthware™ Claisen Adapter with 45° Side Arm
GREENER_CHOICESDP
Catalog No. 31500687 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
24/40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-687 14/20
31-500-688 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-500-687 Supplier Synthware™ Supplier No. A111420
Add to Cart
Add to Cart
Claisen adapter with an additional standard taper outer joint at a 45 deg. angle to the vertical outer joint
  • Three-inlet type Claisen adapter for multiple entries into a single neck flask
  • One side joint is at a 45° angle
  • All joints are the same size
Specifications
Material Borosilicate Glass (3.3)
Product Type Claisen Adapter
Standard Taper Joint No. 14/20