Learn More
Description
- Upper outer joint and lower inner joint are of the same size.
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Material | Glass |
| Standard Taper Joint No | 24/40 |
| Type | Coil Condenser |
| Diameter (Metric) Outer Tubulation | 10 mm |
| Length (Metric) Jacket | 400 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

