Learn More
Synthware™ Connecting Adapter
GREENER_CHOICESDP
Catalog No. 31500689 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
24/40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-689 14/20
31-500-690 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-500-689 Supplier Synthware™ Supplier No. A101420
Add to Cart
Add to Cart
Three-arm glass joint adapters for the assembly of three vessels
Synthware connecting adapter is a 3-way adapter used for connecting three vessels. It has one top outer joint and two lower inner joints, all being the same size.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Glass Joint Adapter
Quantity 1
Standard Taper Joint No. 14/20