Learn More
Synthware™ Connecting Adapters
GREENER_CHOICESDP
Catalog No. 31500685 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
24/40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-685 14/20
31-500-686 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-500-685 Supplier Synthware™ Supplier No. A331420
Add to Cart
Add to Cart
Three-arm design for the assembly of three vessels
Synthware connecting adapter is a 3-way adapter used for connecting three vessels. It has two top outer joints and a lower inner joint, all being the same size.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Connecting adapter
Quantity 1
Standard Taper Joint No. 14/20