Learn More
Synthware™ Distillation Adapter, 75°
GREENER_CHOICESDP
Catalog No. 31500691 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-691 14/20
31-500-692 19/22
31-500-693 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-691 Supplier Synthware™ Supplier No. A151420
Add to Cart
Add to Cart
Distillation adapter with standard taper inner joints on each end at a 75 deg. angle
Synthware distillation adapter bent at 75 degrees. Both ends are standard taper inner joints and are the same size
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Glass Joint Adapter
Quantity 1
Standard Taper Joint No. 14/20